missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lysine 97.0+%, TCI America™

Catalog No. L01295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01295G 5 g
L012925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01295G Supplier TCI America Supplier No. L01295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-87-1 | C6H14N2O2 | 146.19 g/mol

Chemical Identifiers

CAS 56-87-1
Molecular Formula C6H14N2O2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00064433
InChI Key KDXKERNSBIXSRK-MLHKIVSYNA-N
Synonym l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum
PubChem CID 5962
ChEBI CHEBI:18019
IUPAC Name (2S)-2,6-diaminohexanoic acid
SMILES NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 5 g
Formula Weight 146.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-(+)-Lysine
EINECSNumber (9)-1633
RTECSNumber OL5540000
TSCA Yes