CAS RN 559-70-6
CAS RN 559-70-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
β-Amyrin, ≥98.5% (HPLC), MilliporeSigma™ Supelco™
β-Amyrins are pentacyclic triterpenes and a large class of secondary metabolites, isolated from various plants. It exhibits gastroprotective, antipruritic and hepatoprotective activities.
2
phyproof™ β-Amyrin, ≥90% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG