missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ β-Amyrin, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 559-70-6 |
| Product Line | phyproof |
| Linear Formula | C30H50 O |
| MDL Number | MFCD00017381 |
| Synonym | beta-Amyrenol; Olean-12-en-3 beta-ol |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 426.72 |
| Percent Purity | ≥90% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG