accessibility menu, dialog, popup

UserName

(S)OHO(R)HNH3COHOOCH3CH3CH3

CAS RN 55674-67-4

IUPAC Name: (2R,3S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxybutanoic acid
Synonyms: (2R,3S)-2-[(tert-butoxycarbonyl)amino]-3-hydroxybutanoic acid|BOC-D-THR-OH
Molecular Weight (g/mol): 219.24
Molecular Formula: C9H17NO5
InChi Key: LLHOYOCAAURYRL-NTSWFWBYSA-N
SMILES: C[C@H](O)[C@@H](NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)