CAS RN 556109-99-0
CAS RN 556109-99-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2-Morpholinopyrid-4-yl)methanol, 97%, Thermo Scientific™
CAS: 556109-99-0 Molecular Formula: C10H14N2O2 Molecular Weight (g/mol): 194.234 MDL Number: MFCD08271915 InChI Key: ODPRUQYSXQSWQA-UHFFFAOYSA-N Synonym: 2-morpholinopyrid-4-yl methanol,2-morpholin-4-yl pyridin-4-yl methanol,2-morpholin-4-ylpyridin-4-yl methanol,2-morpholinopyridin-4-yl methanol,2-morpholin-4-yl-4-pyridyl methan-1-ol,n-4-hydroxymethylpyridin-2-yl morpholine,4-hydroxymethyl-2-morpholin-4-yl pyridine PubChem CID: 7537566 IUPAC Name: (2-morpholin-4-ylpyridin-4-yl)methanol SMILES: C1COCCN1C2=NC=CC(=C2)CO