missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholinopyrid-4-yl)methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 556109-99-0
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.234
MDL Number MFCD08271915
InChI Key ODPRUQYSXQSWQA-UHFFFAOYSA-N
Synonym 2-morpholinopyrid-4-yl methanol, 2-morpholin-4-yl pyridin-4-yl methanol, 2-morpholin-4-ylpyridin-4-yl methanol, 2-morpholinopyridin-4-yl methanol, 2-morpholin-4-yl-4-pyridyl methan-1-ol, n-4-hydroxymethylpyridin-2-yl morpholine, 4-hydroxymethyl-2-morpholin-4-yl pyridine
PubChem CID 7537566
IUPAC Name (2-morpholin-4-ylpyridin-4-yl)methanol
SMILES C1COCCN1C2=NC=CC(=C2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC43109CB
SDS View Documents
Thermo Scientific Chemicals
CC43109CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC43109DA
SDS View Documents
Thermo Scientific Chemicals
CC43109DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

556109-99-0
194.234
ODPRUQYSXQSWQA-UHFFFAOYSA-N
7537566
C1COCCN1C2=NC=CC(=C2)CO
C10H14N2O2
MFCD08271915
2-morpholinopyrid-4-yl methanol, 2-morpholin-4-yl pyridin-4-yl methanol, 2-morpholin-4-ylpyridin-4-yl methanol, 2-morpholinopyridin-4-yl methanol, 2-morpholin-4-yl-4-pyridyl methan-1-ol, n-4-hydroxymethylpyridin-2-yl morpholine, 4-hydroxymethyl-2-morpholin-4-yl pyridine
(2-morpholin-4-ylpyridin-4-yl)methanol

Specifications

556109-99-0
100.0
Amber Glass Bottle
MFCD08271915
2-morpholinopyrid-4-yl methanol, 2-morpholin-4-yl pyridin-4-yl methanol, 2-morpholin-4-ylpyridin-4-yl methanol, 2-morpholinopyridin-4-yl methanol, 2-morpholin-4-yl-4-pyridyl methan-1-ol, n-4-hydroxymethylpyridin-2-yl morpholine, 4-hydroxymethyl-2-morpholin-4-yl pyridine
C1COCCN1C2=NC=CC(=C2)CO
194.234
194.23
(2-Morpholinopyrid-4-yl)methanol
96.5
97%
C10H14N2O2
250mg
ODPRUQYSXQSWQA-UHFFFAOYSA-N
(2-morpholin-4-ylpyridin-4-yl)methanol
7537566
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'