missing translation for 'onlineSavingsMsg'
Learn More

Allyl iodide, 97%, stabilized

Catalog No. AC218541000
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218541000 100 mL Glass bottle
AC218540250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC218541000 Supplier Thermo Scientific Chemicals Supplier No. 218541000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 556-56-9 | C3H5I | 167.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 556-56-9
Molecular Formula C3H5I
Molecular Weight (g/mol) 167.98
MDL Number MFCD00001094
InChI Key HFEHLDPGIKPNKL-UHFFFAOYSA-N
Synonym allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene
PubChem CID 11166
IUPAC Name 3-iodoprop-1-ene
SMILES ICC=C

Specifications

Melting Point -99.0°C
Color Brown-Red or Orange-Red to Yellow
Density 1.8300g/mL
Boiling Point 102.0°C to 103.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5530 to 1.5550
Linear Formula H2C=CHCH2I
Quantity 100 mL
Beilstein 01,202
Specific Gravity 1.83
Merck Index 15,285
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
Stabilizer Copper
Formula Weight 167.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Allyl iodide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 209-130-4

RUO – Research Use Only