accessibility menu, dialog, popup

UserName

AlH3COH3COH3CO

CAS RN 555-75-9

IUPAC Name: aluminium(3+) triethanolate
Synonyms: aluminium triethoxide
Molecular Weight (g/mol): 162.17
Molecular Formula: C6H15AlO3
InChi Key: JPUHCPXFQIXLMW-UHFFFAOYSA-N
SMILES: [Al+3].CC[O-].CC[O-].CC[O-]
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)