missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum ethoxide
Description
Aluminum ethoxide is mainly used in industrial settings. It is used as a reducing agent for aldehyde, ketones, polymerization catalyst. Used in the sol-gel process preparation of high purity aluminum sesquioxide. It also used as a reducing reagent, for carbonyl compounds that restore to alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Melting Point | 154°C to 159°C |
| Color | White |
| Density | 1.142 g/mL |
| Boiling Point | 320°C |
| Linear Formula | Al(OC2H5)3 |
| Quantity | 250 g |
| UN Number | UN2925 |
| Sensitivity | Moisture sensitive |
| Merck Index | 14,338 |
| Solubility Information | Soluble in hot xylene,cholorobenzene. Reacts with water. |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H228-H261-H314 |
| Hazard Statement | GHS H Statement H228-H314 Flammable solid. Causes severe skin burns and eye damage. |
| Precautionary Statement | P210-P223-P231+P232-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 4.3; Packaging Group: II |
| EINECSNumber | 209-105-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only