missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Ethoxide 98.0+%, TCI America™

Catalog No. A024525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A024525G 25 g
A0245250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A024525G Supplier TCI America Supplier No. A024525G
Only null left

Chemical Identifiers

CAS 555-75-9
Molecular Formula C6H15AlO3-3
Molecular Weight (g/mol) 162.165
MDL Number MFCD00009658
InChI Key JBRFPFQROJYWFN-UHFFFAOYSA-N
Synonym acmc-209lnt
PubChem CID 70700059
IUPAC Name aluminum;ethanolate
SMILES CC[O-].CC[O-].CC[O-].[Al]

Specifications

Melting Point 140°C
Color White-Yellow
Boiling Point 200°C
Quantity 25 g
UN Number 2925
Formula Weight 162.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aluminum Ethoxide
EINECSNumber (2)-2388
TSCA Yes