CAS RN 554-01-8
IUPAC Name:
6-amino-5-methyl-1,2-dihydropyrimidin-2-one
Synonyms:
5-methylcytosine
Molecular Weight (g/mol):
125.13
Molecular Formula:
C5H7N3O
InChi Key:
LRSASMSXMSNRBT-UHFFFAOYSA-N
SMILES:
CC1=C(N)NC(=O)N=C1
1
–
1
of
1
results
1
Promotions Available
5-Methylcytosine, 97%
5-Methylcytosine, 97%
CAS: 554-01-8 Molecular Formula: C5H7N3O Molecular Weight (g/mol): 125.13 MDL Number: MFCD00233537 InChI Key: LRSASMSXMSNRBT-UHFFFAOYSA-N Synonym: 5-methylcytosine,cytosine, 5-methyl,2 1h-pyrimidinone, 4-amino-5-methyl,5-methylcytosine van,5 methylcytosine,5-methyl-cytosine,unii-6r795cqt4h,4-amino-5-methyl-2-pyrimidinol,4-amino-5-methylpyrimidin-2 1h-one,cytosine, 5-methyl-van PubChem CID: 65040 ChEBI: CHEBI:27551 IUPAC Name: 6-amino-5-methyl-1H-pyrimidin-2-one SMILES: CC1=C(N)NC(=O)N=C1