missing translation for 'onlineSavingsMsg'
Learn More

5-Methylcytosine, 97%

Catalog No. AA43492ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43492ME 500 mg
AA43492MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43492ME Supplier Thermo Scientific Chemicals Supplier No. 043492ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 554-01-8 | C5H7N3O | 125.13 g/mol

5-Methylcytosine is used as a marker to protect DNA from being cut by native methylation-sensitive restriction enzymes. It is also employed in the production of 4-amino-5-hydroxy-5-methyl-2-oxo-2,5-dihydro-imidazole-1-carbaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 554-01-8
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.13
MDL Number MFCD00233537
InChI Key LRSASMSXMSNRBT-UHFFFAOYSA-N
Synonym 5-methylcytosine, cytosine, 5-methyl, 2 1h-pyrimidinone, 4-amino-5-methyl, 5-methylcytosine van, 5 methylcytosine, 5-methyl-cytosine, unii-6r795cqt4h, 4-amino-5-methyl-2-pyrimidinol, 4-amino-5-methylpyrimidin-2 1h-one, cytosine, 5-methyl-van
PubChem CID 65040
ChEBI CHEBI:27551
IUPAC Name 6-amino-5-methyl-1H-pyrimidin-2-one
SMILES CC1=C(N)NC(=O)N=C1

Specifications

Quantity 500 mg
Merck Index 14,6050
Solubility Information Soluble in water.
Formula Weight 125.13
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 5-Methylcytosine
EINECSNumber 209-058-3
RTECSNumber UW7362350
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only