accessibility menu, dialog, popup

UserName

NH2ClCl

CAS RN 554-00-7

IUPAC Name: 2,4-dichloroaniline
Synonyms: 2,4-dichloroaniline
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: KQCMTOWTPBNWDB-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1Cl
Boiling Point 
  • (10)
Molecular Weight (g/mol) 
  • (7)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (7)
  • (10)
  • (7)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (6)