CAS RN 552330-86-6
IUPAC Name:
5-bromo-2,3-dihydro-1H-isoindol-1-one
Synonyms:
5-bromo-2,3-dihydroisoindol-1-one
Molecular Weight (g/mol):
212.05
Molecular Formula:
C8H6BrNO
InChi Key:
WJNKJYJCWXMBNV-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)C(=O)NC2
1
–
1
of
1
results
1
5-Bromoisoindolin-1-one 98.0+%, TCI America™
CAS: 552330-86-6 Molecular Formula: C8H6BrNO Molecular Weight (g/mol): 212.046 MDL Number: MFCD09701292 InChI Key: WJNKJYJCWXMBNV-UHFFFAOYSA-N Synonym: 5-bromoisoindolin-1-one,5-bromo-2,3-dihydro-1h-isoindol-1-one,5-bromo-2,3-dihydro-isoindol-1-one,5-bromo-1-isoindolinone,1h-isoindol-1-one, 5-bromo-2,3-dihydro,5-bromoisoindolinone,acmc-1aw2c,5-bromo-1-oxo-isoindoline,5-bromo-2,3-dihydro-1h-isoindole-1-one PubChem CID: 22607119 IUPAC Name: 5-bromo-2,3-dihydroisoindol-1-one SMILES: C1C2=C(C=CC(=C2)Br)C(=O)N1