missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoisoindolin-1-one 98.0+%, TCI America™

Catalog No. B53281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B53281G Supplier TCI America Supplier No. B53281G
Only null left

Chemical Identifiers

CAS 552330-86-6
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
MDL Number MFCD09701292
InChI Key WJNKJYJCWXMBNV-UHFFFAOYSA-N
Synonym 5-bromoisoindolin-1-one, 5-bromo-2,3-dihydro-1h-isoindol-1-one, 5-bromo-2,3-dihydro-isoindol-1-one, 5-bromo-1-isoindolinone, 1h-isoindol-1-one, 5-bromo-2,3-dihydro, 5-bromoisoindolinone, acmc-1aw2c, 5-bromo-1-oxo-isoindoline, 5-bromo-2,3-dihydro-1h-isoindole-1-one
PubChem CID 22607119
IUPAC Name 5-bromo-2,3-dihydroisoindol-1-one
SMILES C1C2=C(C=CC(=C2)Br)C(=O)N1

Specifications

Melting Point 267°C
Quantity 1 g
Formula Weight 212.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoisoindolin-1-one