missing translation for 'onlineSavingsMsg'
Learn More

Methyl pivaloylacetate, 98%, Thermo Scientific™

Catalog No. p-4259033
Change view
Click to view available options
Quantity:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC414190010 1g
AC414190100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414190010 Supplier Thermo Scientific Chemicals Supplier No. 414190010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 55107-14-7
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.2
InChI Key XTXCFTMJPRXBBC-UHFFFAOYSA-N
Synonym methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2
PubChem CID 99597
IUPAC Name methyl 4,4-dimethyl-3-oxopentanoate
SMILES CC(C)(C)C(=O)CC(=O)OC

Specifications

Density 0.9800g/mL
Boiling Point 67.0°C to 70.0°C (13.0mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4300 to 1.4320
Linear Formula (CH3)3CCOCH2COOCH3
Quantity 1g
Specific Gravity 0.98
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material Methyl pivaloylacetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 259-481-2

RUO – Research Use Only