accessibility menu, dialog, popup

UserName

CAS RN 54881-13-9

IUPAC Name: 1-benzylazetidin-3-ol
Synonyms: 1-benzylazetidin-3-ol
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: JOXQHYFVXZZGQZ-UHFFFAOYSA-N
SMILES: OC1CN(CC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results