accessibility menu, dialog, popup

UserName

CAS RN 548-83-4

HOOHOOOH

CAS RN 548-83-4

IUPAC Name: 3,5,7-trihydroxy-2-phenyl-4H-chromen-4-one
Synonyms: galangin 3,5,7-triOH-flavone norizalpinin
Molecular Weight (g/mol): 270.24
Molecular Formula: C15H10O5
InChi Key: VCCRNZQBSJXYJD-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=CC=C2)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results