missing translation for 'onlineSavingsMsg'
Learn More

Galangin, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001998
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001998 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001998 Supplier Merck Emd Millipore Supplier No. 9234225MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Galangin is a polyphenolic compound belonging to the class of flavonoids and is primarily derived from different medicinal herbs. It exhibits antiproliferative, antiangiogenic, antioxidant, anti-cancer, antiinflammatory effects and muscle contraction inhibitor properties.

Featured Industries: Food and Beverages

Specifications

CAS 548-83-4
Physical Form Powder/Crystals
Linear Formula C15H10O5
MDL Number MFCD00006833
Melting Point 214°C to 215°C
Synonym 3,5,7-Trihydroxyflavone; Norizalpinin
Type Analytical Standard
Quantity 25 mg
Formula Weight 270.24
Percent Purity ≥97% (HPLC)
Grade Analytical Standard
Show More Show Less