accessibility menu, dialog, popup

UserName

CAS RN 547-91-1

NaOHOOOHINSOHOO

CAS RN 547-91-1

IUPAC Name: sodium 8-hydroxy-7-iodoquinoline-5-sulfonic acid hydrogen carbonate
Synonyms: chiniofon
Molecular Weight (g/mol): 435.12
Molecular Formula: C10H7INNaO7S
InChi Key: FNXKBSAUKFCXIK-UHFFFAOYSA-M
SMILES: [Na+].OC([O-])=O.OC1=C2N=CC=CC2=C(C=C1I)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)