missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-7-iodoquinoline-5-sulfonic Acid 98.0+%, TCI America™

Catalog No. H025425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H025425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H025425G Supplier TCI America Supplier No. H025425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 547-91-1
Molecular Formula C9H6INO4S
Molecular Weight (g/mol) 351.114
MDL Number MFCD00006793
InChI Key ZBJWWKFMHOAPNS-UHFFFAOYSA-N
Synonym ferron, loretin, quiniophen, meditrene, anayodin, quinoxyl, sefona, yatren, yellon, 8-hydroxy-7-iodo-5-quinolinesulfonic acid
PubChem CID 11043
IUPAC Name 8-hydroxy-7-iodoquinoline-5-sulfonic acid
SMILES C1=CC2=C(C(=C(C=C2S(=O)(=O)O)I)O)N=C1

Specifications

Melting Point 270°C
Color Yellow
Quantity 25 g
UN Number 3261
Formula Weight 351.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Hydroxy-7-iodoquinoline-5-sulfonic Acid
RTECSNumber VC2800000
TSCA Yes