accessibility menu, dialog, popup

UserName

CAS RN 5460-29-7

IUPAC Name: 2-(3-bromopropyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: N-(3-bromopropyl)phthalimide
Molecular Weight (g/mol): 268.11
Molecular Formula: C11H10BrNO2
InChi Key: VKJCJJYNVIYVQR-UHFFFAOYSA-N
SMILES: BrCCCN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)