accessibility menu, dialog, popup

UserName

NNHBrHBr

CAS RN 54581-69-0

IUPAC Name: 1,4-diazabicyclo[2.2.2]octane dihydrobromide
Synonyms: dabco dihydrobromide
Molecular Weight (g/mol): 274
Molecular Formula: C6H14Br2N2
InChi Key: GPELLQBNVCIHJJ-UHFFFAOYSA-N
SMILES: Br.Br.C1CN2CCN1CC2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results