missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diazabicyclo[2.2.2]octane Dihydrobromide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 319°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 274.00 |
| Percent Purity | ≥98.0% (N,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,4-Diazabicyclo[2.2.2]octane Dihydrobromide |
Safety and Handling
| TSCA | No |