accessibility menu, dialog, popup

UserName

CH3ONHNHO

CAS RN 5444-02-0

IUPAC Name: 2-hydroxy-4-methyl-6-oxo-1,6-dihydropyridine-3-carbonitrile
Synonyms: 2-hydroxy-4-methyl-6-oxo-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol): 150.14
Molecular Formula: C7H6N2O2
InChi Key: YRGYYQCOWUULNF-UHFFFAOYSA-N
SMILES: CC1=CC(=O)NC(O)=C1C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results