missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyano-6-hydroxy-4-methyl-2-pyridone 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 300°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 150.14 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Cyano-6-hydroxy-4-methyl-2-pyridone |
Safety and Handling
| TSCA | Yes |