accessibility menu, dialog, popup

UserName

CH3OHONOO

CAS RN 5437-38-7

IUPAC Name: 3-methyl-2-nitrobenzoic acid
Synonyms: 3-methyl-2-nitrobenzoic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: DGDAVTPQCQXLGU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(O)=O)=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)