CAS RN 5411-56-3
IUPAC Name:
1-(2-bromophenyl)ethan-1-ol
Synonyms:
1-(2-bromophenyl)ethanol
Molecular Weight (g/mol):
201.06
Molecular Formula:
C8H9BrO
InChi Key:
DZLZSFZSPIUINR-UHFFFAOYNA-N
SMILES:
CC(O)C1=CC=CC=C1Br
1
–
1
of
1
results
1
Promotions Available
2-Bromo-alpha-methylbenzyl alcohol, 99%
2-Bromo-alpha-methylbenzyl alcohol, 99%
CAS: 5411-56-3 Molecular Formula: C8H9BrO Molecular Weight (g/mol): 201.06 MDL Number: MFCD00065001 InChI Key: DZLZSFZSPIUINR-UHFFFAOYSA-N Synonym: 1-2-bromophenyl ethanol,2-bromo-alpha-methylbenzyl alcohol,1-2-bromophenyl ethan-1-ol,1-2'-bromophenyl-1-hydroxyethane,acmc-209wki,o-bromo-1-phenylethanol,o-bromo-1-phenyl ethanol,acmc-209y6n,1-2-bro-mophenyl ethanol,1-2-bromo-phenyl-ethanol PubChem CID: 95455 IUPAC Name: 1-(2-bromophenyl)ethanol SMILES: CC(C1=CC=CC=C1Br)O