accessibility menu, dialog, popup

UserName

OBrFFF

CAS RN 54109-16-9

IUPAC Name: 2-bromo-1-[2-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[2-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 267.05
Molecular Formula: C9H6BrF3O
InChi Key: KWZCBMKXNYOQAK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1C(=O)CBr
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)