missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-(trifluoromethyl)acetophenone 98.0+%, TCI America™

Catalog No. B47971G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47971G Supplier TCI America Supplier No. B47971G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54109-16-9
Molecular Formula C9H6BrF3O
Molecular Weight (g/mol) 267.045
MDL Number MFCD03094304
InChI Key KWZCBMKXNYOQAK-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenacyl bromide, 2-bromo-1-2-trifluoromethyl phenyl-1-ethanone, 2-bromo-2'-trifluoromethyl acetophenone, 2-bromo-1-2-trifluoromethyl-phenyl-ethanone, 2-bromo-1-2-trifluoromethyl phenyl ethan-1-one, 2-bromo-1-2-trifluoromethyl phenyl ethanone, ethanone, 2-bromo-1-2-trifluoromethyl phenyl, 2-trifluoromethyphenacyl bromide, 2-trifluoromethyl phenacylbromide
PubChem CID 2778430
IUPAC Name 2-bromo-1-[2-(trifluoromethyl)phenyl]ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)C(F)(F)F

Specifications

Melting Point 30°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 267.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-2′-(trifluoromethyl)acetophenone
TSCA No
Recommended Storage Refrigerator