missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-(trifluoromethyl)acetophenone, 97%

Catalog No. AAH3203206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3203206 5 g
AAH3203203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3203206 Supplier Thermo Scientific Chemicals Supplier No. H3203206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54109-16-9 | C9H6BrF3O | 267.045 g/mol

The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 54109-16-9
Molecular Formula C9H6BrF3O
Molecular Weight (g/mol) 267.045
MDL Number MFCD03094304
InChI Key KWZCBMKXNYOQAK-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenacyl bromide, 2-bromo-1-2-trifluoromethyl phenyl-1-ethanone, 2-bromo-2'-trifluoromethyl acetophenone, 2-bromo-1-2-trifluoromethyl-phenyl-ethanone, 2-bromo-1-2-trifluoromethyl phenyl ethan-1-one, 2-bromo-1-2-trifluoromethyl phenyl ethanone, ethanone, 2-bromo-1-2-trifluoromethyl phenyl, 2-trifluoromethyphenacyl bromide, 2-trifluoromethyl phenacylbromide
PubChem CID 2778430
IUPAC Name 2-bromo-1-[2-(trifluoromethyl)phenyl]ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)C(F)(F)F

Specifications

Melting Point 24°C to 32°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 267.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-2'-(trifluoromethyl)acetophenone
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only