accessibility menu, dialog, popup

UserName

CH3NH2O

CAS RN 541-35-5

IUPAC Name: butanamide
Synonyms: butyramide N-butyramide C3H7C(O)NH2 butyrylamide amide butanoic acid n-C3H7C(O)NH2
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: DNSISZSEWVHGLH-UHFFFAOYSA-N
SMILES: CCCC(N)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)