missing translation for 'onlineSavingsMsg'
Learn More

Butyramide, 98%

Catalog No. AAA1249436
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1249436 500 g
AAA1249418 50 g
AAA1249422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1249436 Supplier Thermo Scientific Chemicals Supplier No. A1249436

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 541-35-5 | C4H9NO | 87.122 g/mol

Butyramide was used in the synthesis of hydroxamic acids, electrorheological fluids and β-amodoorganotin compounds. It was used as substrate of (+)-γ-lactamase to develop a microreactor to study enzyme stability, activity, kinetics and substrate specificity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyramide was used in the synthesis of hydroxamic acids, electrorheological fluids and β-amodoorganotin compounds. It was used as substrate of (+)-γ-lactamase to develop a microreactor to study enzyme stability, activity, kinetics and substrate specificity.

Solubility
Soluble in water (170 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 541-35-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00041894
InChI Key DNSISZSEWVHGLH-UHFFFAOYSA-N
Synonym butyramide, n-butyramide, n-butylamide, butanimidic acid, butanoic acid, amide, unii-9j6or937vr, butyroamide, n-c3h7c o nh2, butyrylamide, butyramid
PubChem CID 10927
ChEBI CHEBI:50724
IUPAC Name butanamide
SMILES CCCC(=O)N

Specifications

Melting Point 113°C to 116°C
Density 1.032
Boiling Point 216°C
Quantity 500 g
Beilstein 1361528
Merck Index 14,1592
Solubility Information Soluble in water (170g/L).
Formula Weight 87.12
Percent Purity 98%
Chemical Name or Material Butyramide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 208-776-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only