accessibility menu, dialog, popup

UserName

CAS RN 54024-22-5

CH3(S)CH2(S)(S)(R)(S)(R)HOHC

CAS RN 54024-22-5

IUPAC Name: (1R,3aS,3bS,9aR,9bS,11aS)-11a-ethyl-1-ethynyl-10-methylidene-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol
Synonyms: desogestrel desogestrel product organon brand of desogestrel cerazette desogestrel preparation desogestrel preparation (product) desogestrel (substance) mircette desogen desogestrelum
Molecular Weight (g/mol): 310.48
Molecular Formula: C22H30O
InChi Key: RPLCPCMSCLEKRS-BPIQYHPVSA-N
SMILES: CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results