missing translation for 'onlineSavingsMsg'
Learn More

Isoniazid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002000 Supplier Merck Emd Millipore Supplier No. PHR1932500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 54-85-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C6H7N3 O
MDL Number MFCD00006426
Melting Point 171°C to 173°C
Synonym Isoniazid; 4-Pyridinecarboxylic acid hydrazide; INH; Isonicotinic acid hydrazide; Isonicotinic hydrazide
Type Certified Reference Material
Quantity 500 mg
Formula Weight 137.14
Grade Certified Reference Material