accessibility menu, dialog, popup

UserName

CAS RN 53936-56-4

IUPAC Name: 4-(oxan-2-yloxy)phenol
Synonyms: tetrahydropyranyloxy phenol
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: GFBCWCDNXDKFRH-UHFFFAOYNA-N
SMILES: OC1=CC=C(OC2CCCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results