accessibility menu, dialog, popup

UserName

CAS RN 538-28-3

IUPAC Name: hydrogen (benzylsulfanyl)methanimidamide chloride
Synonyms: hydrogen carbamimidothioic acid, phenylmethyl ester (9CI) chloride
Molecular Weight (g/mol): 202.7
Molecular Formula: C8H11ClN2S
InChi Key: WJAASTDRAAMYNK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(=N)SCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results