accessibility menu, dialog, popup

UserName

H3CN

CAS RN 536-75-4

IUPAC Name: 4-ethylpyridine
Synonyms: 4-ethylpyridine
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: VJXRKZJMGVSXPX-UHFFFAOYSA-N
SMILES: CCC1=CC=NC=C1
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (5)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)