missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylpyridine 98.0+%, TCI America™

Catalog No. E017025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E017025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E017025ML Supplier TCI America Supplier No. E017025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 536-75-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006450
InChI Key VJXRKZJMGVSXPX-UHFFFAOYSA-N
Synonym pyridine, 4-ethyl, .gamma.-ethylpyridine, 4-ethyl pyridine, unii-3kw0se2tl5, 4-ethyl-pyridine, pyridine, c1-3-alkyl derivs., 3kw0se2tl5, gamma-ethylpyridine, c1-c3 alkylpyridine, 4ethylpyridine
PubChem CID 10822
IUPAC Name 4-ethylpyridine
SMILES CCC1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 mL
UN Number 1993
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylpyridine
EINECSNumber (5)-3683&(9)-0222
TSCA Yes