missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylpyridine, 98%
Description
4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.
Solubility
Slightly soluble in water (34 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents and strong acids.
4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.
Solubility
Slightly soluble in water (34 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Melting Point | -91°C |
| Density | 0.942 |
| pH | 10 |
| Boiling Point | 167°C to 168°C |
| Flash Point | 52°C (126°F) |
| Odor | Unpleasant |
| Refractive Index | 1.501 |
| Quantity | 250 g |
| UN Number | UN1993 |
| Beilstein | 106481 |
| Show More |
Safety and Handling
| Hazard Category | H226-H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H StatementH226-H302-H312-H332-H315-H319-H335Flammable liquid and vapour.Harmful if swallowed.Harmful in contact with skin.Harmful if inhaled.Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 208-646-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only