accessibility menu, dialog, popup

UserName

HOBr

CAS RN 53463-68-6

IUPAC Name: 10-bromodecan-1-ol
Synonyms: 10-bromodecan-1-ol
Molecular Weight (g/mol): 237.18
Molecular Formula: C10H21BrO
InChi Key: LGZMUUBPTDRQQM-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCBr
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)