missing translation for 'onlineSavingsMsg'
Learn More

10-Bromo-1-decanol 85.0+%, TCI America™

Catalog No. B17305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17305G 5 g
B173025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17305G Supplier TCI America Supplier No. B17305G

Chemical Identifiers

CAS 53463-68-6
Molecular Formula C10H21BrO
Molecular Weight (g/mol) 237.18
MDL Number MFCD00041681
InChI Key LGZMUUBPTDRQQM-UHFFFAOYSA-N
Synonym 10-bromo-1-decanol, 10-bromodecanol, 1-decanol, 10-bromo, 10-hydroxy-1-bromo decane, 1-bromodecan-10-ol, 10-bromodecane-1-ol, 10-bromo-decan-1-ol, decamethylene bromohydrin, 10-hydroxy1-bromo decane, acmc-209l7p
PubChem CID 104507
IUPAC Name 10-bromodecan-1-ol
SMILES OCCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 169°C
Quantity 5 g
Formula Weight 237.18
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 10-Bromo-1-decanol
TSCA Yes
Recommended Storage Freezer