accessibility menu, dialog, popup

UserName

CAS RN 5345-47-1

NH2NHOO

CAS RN 5345-47-1

IUPAC Name: 2-aminopyridine-3-carboxylic acid
Synonyms: 2-aminopyridine-3-carboxylic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: KPIVDNYJNOPGBE-UHFFFAOYSA-N
SMILES: NC1=NC=CC=C1C(O)=O
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)