missing translation for 'onlineSavingsMsg'
Learn More

2-Aminonicotinic acid, 98+%

Catalog No. AAA1119522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1119522 100 g
AAA1119506 5 g
AAA1119514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1119522 Supplier Thermo Scientific Chemicals Supplier No. A1119522
Only null left

CAS: 5345-47-1 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5345-47-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00006318
InChI Key KPIVDNYJNOPGBE-UHFFFAOYSA-N
Synonym 2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid
PubChem CID 72930
ChEBI CHEBI:68572
IUPAC Name 2-aminopyridine-3-carboxylic acid
SMILES NC1=NC=CC=C1C(O)=O

Specifications

Melting Point ∼298°C (decomposition)
Quantity 100 g
Beilstein 119031
Formula Weight 138.13
Percent Purity ≥98%
Chemical Name or Material 2-Aminonicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-296-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only