missing translation for 'onlineSavingsMsg'
Learn More

2-Aminonicotinic Acid 98.0+%, TCI America™

Catalog No. A09945G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09945G 5 g
A099425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09945G Supplier TCI America Supplier No. A09945G
Only null left

Chemical Identifiers

CAS 5345-47-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00006318
InChI Key KPIVDNYJNOPGBE-UHFFFAOYSA-N
Synonym 2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid
PubChem CID 72930
ChEBI CHEBI:68572
IUPAC Name 2-aminopyridine-3-carboxylic acid
SMILES NC1=NC=CC=C1C(O)=O

Specifications

Quantity 5 g
Formula Weight 138.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminonicotinic Acid