accessibility menu, dialog, popup

UserName

CAS RN 5345-42-6

H3COH3CNOO

CAS RN 5345-42-6

IUPAC Name: 1-methoxy-3-methyl-2-nitrobenzene
Synonyms: 1-methoxy-3-methyl-2-nitrobenzene
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: MGBRGNWARSQECY-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C)=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results