accessibility menu, dialog, popup

UserName

H3CCH3(R)NH2OH

CAS RN 53448-09-2

IUPAC Name: (2R)-1-hydroxy-4-methylpentan-2-aminium
Synonyms: (2R)-1-hydroxy-4-methylpentan-2-aminium
Molecular Weight (g/mol): 118.2
Molecular Formula: C6H16NO
InChi Key: VPSSPAXIFBTOHY-ZCFIWIBFSA-O
SMILES: CC(C)C[C@@H]([NH3+])CO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)