missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Leucinol, 97%, Thermo Scientific Chemicals

Catalog No. AC413120010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413120010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC413120010 Supplier Thermo Scientific Chemicals Supplier No. 413120010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 53448-09-2 | Molecular Formula: C6H16NO | Molecular Weight: 118.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53448-09-2
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00004734
InChI Key VPSSPAXIFBTOHY-ZCFIWIBFSA-O
Synonym d-leucinol, r-2-amino-4-methylpentan-1-ol, 2r-2-amino-4-methylpentan-1-ol, d---leucinol, chembl71171, r---leucinol, 1-pentanol, 2-amino-4-methyl-, 2r, 2-amino-4-methyl-pentan-1-ol, 2-amino-4-methyl-1-pentanol #
PubChem CID 2724002
IUPAC Name (2R)-2-amino-4-methylpentan-1-ol
SMILES CC(C)C[C@@H]([NH3+])CO

Specifications

Density 0.9070g/mL
Boiling Point 199.0°C
Flash Point 90°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4530
Quantity 1 g
Specific Gravity 0.907
Specific Rotation Condition − 4.00 (20.00°C c=9,ethanol)
Specific Rotation − 4.00
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material D(-)-Leucinol, 97%, (97% e.e.)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only