missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Leucinol 97.0+%, TCI America™

Catalog No. L02365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02365G 5 g
L02361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02365G Supplier TCI America Supplier No. L02365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53448-09-2
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00004734
InChI Key VPSSPAXIFBTOHY-ZCFIWIBFSA-O
Synonym d-leucinol, r-2-amino-4-methylpentan-1-ol, 2r-2-amino-4-methylpentan-1-ol, d---leucinol, chembl71171, r---leucinol, 1-pentanol, 2-amino-4-methyl-, 2r, 2-amino-4-methyl-pentan-1-ol, 2-amino-4-methyl-1-pentanol #
PubChem CID 2724002
IUPAC Name (2R)-1-hydroxy-4-methylpentan-2-aminium
SMILES CC(C)C[C@@H]([NH3+])CO

Specifications

Color Yellow
Boiling Point 194°C
Quantity 5 g
Formula Weight 117.19
Percent Purity ≥97.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material D-(-)-Leucinol