accessibility menu, dialog, popup

UserName

CH3O

CAS RN 534-22-5

IUPAC Name: 2-methylfuran
Synonyms: silvan sylvan
Molecular Weight (g/mol): 82.1
Molecular Formula: C5H6O
InChi Key: VQKFNUFAXTZWDK-UHFFFAOYSA-N
SMILES: CC1=CC=CO1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)